Learn More
Abnova™ Human GGA3 Partial ORF (NP_619525, 246 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Alternative splicing of this gene results in two transcript variants. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_619525 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23163 |
Molecular Weight (g/mol) | 36.74kDa |
Name | GGA3 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | ELMKELFDQCENKRRTLFKLASETEDNDNSLGDILQASDNLSRVINSYKTIIEGQVINGEVATLTLPDSEGNSQCSNQGTLIDLAELDTTNSLSSVLAPA |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.