Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ghrelin/Obestatin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8149
This item is not returnable.
View return policy
Description
Ghrelin/Obestatin Polyclonal specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, Immunohistochemistry.Order Info
The product is shipped at ambient temperature.
Specifications
Ghrelin/Obestatin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, GHRL, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46 | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
51738 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9UBU3 | |
GHRL | |
E. coli-derived recombinant human Ghrelin/Obestatin Gly24-Lys117 (Leu72Met) Accession # Q9UBU3 | |
100 μg | |
Primary | |
Detects human Ghrelin/Obestatin in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant mouse Ghrelin/Obestatin is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction