Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GHRHR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10735210
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10735210 100 μg
MAB107352SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10735210 Supplier R&D Systems Supplier No. MAB107352100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GHRHR Monoclonal antibody specifically detects GHRHR in Human samples. It is validated for Western Blot

Specifications

Antigen GHRHR
Applications Western Blot
Classification Monoclonal
Clone 1037308
Conjugate Unconjugated
Dilution Western Blot 4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q02643
Gene Alias GHRFR, GHRH receptor, GRF receptor, GRFR, growth hormone releasing hormone receptor, Growth hormone-releasing factor receptor, growth hormone-releasing hormone receptor, IGHD1B, Little
Host Species Mouse
Immunogen Human GHRHR synthetic peptide, Accession # Q02643
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2692
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.