Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GILT/IFI30 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7715
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7715 Supplier R&D Systems Supplier No. MAB7715
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GILT/IFI30 Monoclonal specifically detects GILT/IFI30 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen GILT/IFI30
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 844623
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13284
Gene Alias EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI30, IFI-30, interferon, gamma-inducible protein 30, IP30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056
Gene Symbols IFI30
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GILT/IFI30 Ser27-Lys250 Accession # P13284
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10437
Test Specificity Detects human GILT/IFI30 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.