Learn More
Human GIPR Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB8210N100UG
Catalog No. FAB8210N100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GIPR
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P48546
GIPR
NS0 mouse myeloma cell line transfected with human GIPR. Met1-Cys466 Accession # P48546
RUO
2696
Human
IgG1
Flow Cytometry
591853
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Mouse
100 μg
Primary
Detects human GIPR in direct ELISAs. Stains human GIPR transfected cells but not irrelevant transfectants.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)