Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GIT1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB85081100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB85081100 100 μg
MAB85081SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB85081100 Supplier R&D Systems Supplier No. MAB85081100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIT1 Monoclonal specifically detects GIT1 in Human samples. It is validated for ELISA.

Specifications

Antigen GIT1
Applications ELISA
Classification Monoclonal
Clone 924616R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q9Y2X7
Gene Alias ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human GIT1 Ser485-Asp636 Accession # Q9Y2X7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 28964
Test Specificity Detects human GIT1 in direct ELISAs. In sandwich ELISAs, this antibody is specific for human GIT1 when paired with the suggested capture antibody.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.