Learn More
Human GITR Ligand/TNFSF18 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | GITR Ligand/TNFSF18 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 109101 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q9UNG2 |
| Gene Alias | Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.