Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GITR Ligand/TNFSF18 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6941G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6941G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6941G100 Supplier R&D Systems Supplier No. FAB6941G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry
Classification Monoclonal
Clone 109101
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UNG2
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbols TNFSF18
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18 Glu52-Ser177 Accession # Q9UNG2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8995
Test Specificity Detects human GITR Ligand/TNFSF18 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.