Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GITR Ligand/TNFSF18 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6941N
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry.Specifications
GITR Ligand/TNFSF18 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Mouse | |
Affinity Purified | |
RUO | |
8995 | |
Human | |
IgG1 |
Flow Cytometry | |
109101 | |
Flow Cytometry 5 ug/mL | |
Q9UNG2 | |
TNFSF18 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18 Glu52-Ser177 Accession # Q9UNG2 | |
100 Tests | |
Primary | |
Detects human GITR Ligand/TNFSF18. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction