Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GITR Ligand/TNFSF18 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6942SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6942SP 25 μg
MAB6942100 100 μg
MAB6942500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB6942SP Supplier R&D Systems Supplier No. MAB6942SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, ELISA
Classification Monoclonal
Clone 109114
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UNG2
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbols TNFSF18
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18 Glu52-Ser177 Accession # Q9UNG2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8995
Test Specificity Detects human GITR Ligand/TNFSF18 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) APRIL, rhLIGHT, rhTNF-alpha , or rhVEGI is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.