Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GITR Ligand/TNFSF18 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $573.00
Specifications
Antigen | GITR Ligand/TNFSF18 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Classification | Polyclonal |
Description
GITR Ligand/TNFSF18 Polyclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
GITR Ligand/TNFSF18 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
TNFSF18 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
RUO | |
Q9UNG2 | |
8995 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18 Glu52-Ser177 Accession # Q9UNG2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human GITR Ligand/TNSF18 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse GITR Ligand is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title