Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GITR/TNFRSF18 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF689SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF689SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF689SP Supplier R&D Systems Supplier No. AF689SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GITR/TNFRSF18 Polyclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready.

Specifications

Antigen GITR/TNFRSF18
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y5U5
Gene Alias Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18
Gene Symbols TNFRSF18
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human GITR/TNFRSF18 Gln26-Glu161 (Thr45Ala) Accession # Q9Y5U5
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8784
Test Specificity Detects human GITR/TNFRSF18 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse GITR and recombiant human 4-1BB is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.