Learn More
Human GITR/TNFRSF18 Fluorescein-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | GITR/TNFRSF18 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 110416 |
| Conjugate | Fluorescein |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q9Y5U5 |
| Gene Alias | Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18 |
| Gene Symbols | TNFRSF18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.