Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GLP-1R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2814
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2814 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2814 Supplier R&D Systems Supplier No. MAB2814
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLP-1R Monoclonal specifically detects GLP-1R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen GLP-1R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 197920
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_002053
Gene Alias GLP-1 receptor, GLP1R, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331
Gene Symbols GLP1R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GLP-1R Arg24-Ser463 Accession # NP_002053
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2740
Test Specificity Stains human GLP-1R transfectants but not irrelevant transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.