Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glucagon R/GCGR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $473.00
Specifications
Antigen | Glucagon R/GCGR |
---|---|
Clone | 591929 |
Dilution | Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10296SP
![]() |
R&D Systems
MAB10296SP |
25 μg |
Each for $208.00
|
|
|||||
MAB10296100
![]() |
R&D Systems
MAB10296100 |
100 μg |
Each for $473.00
|
|
|||||
Description
Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Glucagon R/GCGR | |
Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
FLJ97182, GCGR, GGR, GLR, GL-R, glucagon receptor, GlucagonR, MGC138246 | |
HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Glucagon R/GCGR in direct ELISAs. |
591929 | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P47871 | |
2642 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title