Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GluR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $454.00
Specifications
Antigen | Glutamate Receptor 4 |
---|---|
Clone | 1059705 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB113631SP
![]() |
R&D Systems
MAB113631SP |
25 μg |
Each for $177.10
|
|
|||||
RD10000108
![]() |
R&D Systems
MAB113631100 |
100 μg |
Each for $454.00
|
|
|||||
Description
Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Glutamate Receptor 4 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4 | |
Synthetic peptide, Accession # P48058 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1059705 | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P48058 | |
2893 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title