Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GluR4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$184.70 - $454.00
Specifications
Antigen | Glutamate Receptor 4 |
---|---|
Clone | 2913C |
Dilution | Immunohistochemistry 1-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
RD10000182
![]() |
R&D Systems
MAB113633100 |
100 μg |
Each for $454.00
|
|
|||||
MAB113633SP
![]() |
R&D Systems
MAB113633SP |
25 μg |
Each for $184.70
|
|
|||||
Description
Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Human samples. It is validated for ImmunohistochemistrySpecifications
Glutamate Receptor 4 | |
Immunohistochemistry 1-15 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4 | |
E. coli-derived recombinant human GluR4, Met112-His314, Accession # P48058.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2913C | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P48058.2 | |
2893 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title