Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GluR7/GRIK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $443.50
Specifications
Antigen | GluR7/GRIK3 |
---|---|
Clone | 1059901 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11328100
![]() |
R&D Systems
MAB11328100 |
100 μg |
Each for $443.50
|
|
|||||
MAB11328SP
![]() |
R&D Systems
MAB11328SP |
25 μg |
Each for $177.10
|
|
|||||
Description
GluR7/GRIK3 Monoclonal antibody specifically detects GluR7/GRIK3 in Human samples. It is validated for ImmunohistochemistrySpecifications
GluR7/GRIK3 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
EAA5, Excitatory amino acid receptor 5, GLR7, GluK3, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3, GRIK3 | |
Human GluR7/GRIK3 containing peptide., Accession # Q13003 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1059901 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q13003 | |
2899 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title