Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glut3 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1415S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1415S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1415S100 Supplier R&D Systems Supplier No. FAB1415S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glut3 Monoclonal specifically detects Glut3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glut3
Applications Flow Cytometry
Classification Monoclonal
Clone 202017
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P11169
Gene Alias FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, SLC2A3, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3
Gene Symbols SLC2A3
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Glut3 Accession # P11169
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6515
Test Specificity Detects human Glut3. Recognizes human Glut3 expression on human Glut3-transfected NS0 cells, but not the NS0 control transfectants. No cross-reactivity was observed with transfectants expressing human Glut1 or human Glut2.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.