Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glut4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB86541G100UG
This item is not returnable.
View return policy
Description
Glut4 Monoclonal specifically detects Glut4 in Human samples. It is validated for Flow Cytometry.Specifications
Glut4 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, SLC2A4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
Mouse myeloma cell line NS0-derived recombinant human Glut4 Met1-Asp509 Accession # P14672 | |
100 μg | |
Primary | |
Detects human Glut4 in direct ELISAs. | |
Human | |
IgG2b |
Flow Cytometry | |
925932 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P14672 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6517 | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction