Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glut5 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1349S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1349S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1349S100 Supplier R&D Systems Supplier No. FAB1349S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glut5 Monoclonal specifically detects Glut5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glut5
Applications Flow Cytometry
Classification Monoclonal
Clone 195205
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P22732
Gene Alias Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, SLC2A5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5
Gene Symbols SLC2A5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Glut5 Met1-Gln501 Accession # P22732
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6518
Test Specificity Detects human Glut5. Stains human Glut5 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.