Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glut5 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM1349
This item is not returnable.
View return policy
Description
Glut5 Monoclonal specifically detects Glut5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
Glut5 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL | |
P22732 | |
SLC2A5 | |
NS0 mouse myeloma cell line transfected with human Glut5 Met1-Gln501 Accession # P22732 | |
200 μg | |
Primary | |
Detects human Glut5. Stains human Glut5-transfected NS0 cells; also detects Glut5 on human microglia cells (Sasaki, A. et al. (2003) Neurosci. Lett. 338(1):17). | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry | |
195205 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, SLC2A5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6518 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction