Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Glutamate Dehydrogenase ELISA Kit (Colorimetric)
SDP

Catalog No. NB170746
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.31-20 mIU/mL
Assay Sensitivity 0.19 mIU/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2746
Gene Symbol GLUD1
Quantity 1 Kit
Research Discipline Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 1.4.1, EC 1.4.1.3, GDH, GDH 1, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, glutamate dehydrogenase 1, mitochondrial, MGC132003
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.