Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glutathione S-Transferase pi 1/GSTP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6455SP
This item is not returnable.
View return policy
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Glutathione S-Transferase pi 1/GSTP1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 12.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
AAC51280 | |
GSTP1 | |
E. coli-derived recombinant human Glutathione S-Transferase pi 1/GSTP1 Met1-Glu210 Accession # AAC51280 | |
25 μg | |
Primary | |
Detects human Glutathione S-Transferase pi 1/GSTP1 in direct ELISAs and Western blots. Detects mouse and rat Glutathione S-Transferase pi 1/GSTP1 in Western Blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
800027 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
deafness, X-linked 7, DFN7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, Glutathione STransferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1, GSTP1-1, PI | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2950 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction