Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glutathione S-Transferase pi 1/GSTP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6455
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6455 Supplier R&D Systems Supplier No. AF6455
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Glutathione S-Transferase pi 1/GSTP1 Polyclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Glutathione S-Transferase pi 1/GSTP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. AAC51280
Gene Alias deafness, X-linked 7, DFN7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, Glutathione STransferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1, GSTP1-1, PI
Gene Symbols GSTP1
Host Species Sheep
Immunogen E. coli-derived recombinant human Glutathione S-Transferase pi 1/GSTP1 Met1-Glu210 Accession # AAC51280
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2950
Test Specificity Detects human Glutathione S-Transferase pi 1/GSTP1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.