Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glycoprotein V/CD42d Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4249V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4249V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4249V100 Supplier R&D Systems Supplier No. FAB4249V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glycoprotein V/CD42d
Applications Flow Cytometry
Classification Monoclonal
Clone 508209
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P40197
Gene Alias platelet glycoprotein V, CD42d, glycoprotein V (platelet), GP5, GPV
Gene Symbols GP5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d Gln17-Gly523 Accession # P40197
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2814
Test Specificity Detects human Glycoprotein V/CD42d in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human GPVI is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.