Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glypican 1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4519
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4519 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4519 Supplier R&D Systems Supplier No. BAF4519
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Glypican 1 Polyclonal specifically detects Glypican 1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Glypican 1
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells
Gene Accession No. P35052
Gene Alias FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1, GPC1
Gene Symbols GPC1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glypican 1 Asp24-Ser530 Accession # P35052
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Test Specificity Detects human Glypican 1 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse Glypican 1 is observed and less than 1% cross-reactivity with recombinant human (rh)Glypican 5 and rhGlypican 6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.