Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glypican 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Glypican 2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
Glypican 2 Polyclonal specifically detects Glypican 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
Glypican 2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
2410016G05Rik, Cerebroglycan, GPC2 | |
GPC2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q8N158 | |
221914 | |
Mouse myeloma cell line NS0-derived recombinant human Glypican 2 Pro18-Ser553 Accession # Q8N158 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Glypican 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse Glypican 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title