Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glypican 4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Glypican4 |
---|---|
Clone | 965523 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB91951SP
![]() |
R&D Systems
MAB91951SP |
25 μg |
Each for $200.50
|
|
|||||
MAB91951100
![]() |
R&D Systems
MAB91951100 |
100 μg |
Each for $503.00
|
|
|||||
Description
Glypican 4 Monoclonal specifically detects Glypican 4 in Human samples. It is validated for ELISA.Specifications
Glypican4 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
O75487 | |
2239 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
965523 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
glypican 4, glypican proteoglycan 4, glypican-4, GPC4, K-glypican, K-glypicandJ900E8.1 (glypican 4) | |
Chinese hamster ovary cell line CHO-derived recombinant human Glypican 4 Met1-Ser529 Accession # O75487 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Glypican 4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title