Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glypican 6 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2845V100UG
This item is not returnable.
View return policy
Description
Glypican 6 Monoclonal specifically detects Glypican 6 in Human samples. It is validated for Flow Cytometry.Specifications
Glypican 6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y625 | |
GPC6 | |
Mouse myeloma cell line NS0-derived recombinant human Glypican 6 Asp24-Val527 Accession # Q9Y625 | |
100 μg | |
Primary | |
Detects human Glypican 6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Glypican 2, 3, or 5 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
348701 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
glypican 6, glypican proteoglycan 6, glypican-6, GPC6, MGC126288, OMIMD1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10082 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction