Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Glypican 6 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Glypican 6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
Glypican 6 Polyclonal specifically detects Glypican 6 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Glypican 6 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
glypican 6, glypican proteoglycan 6, glypican-6, GPC6, MGC126288, OMIMD1 | |
GPC6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q9Y625 | |
10082 | |
Mouse myeloma cell line NS0-derived recombinant human Glypican 6 Asp24-Val527 Accession # Q9Y625 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Glypican 6 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title