Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GM-CSF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$214.00 - $2200.00
Specifications
| Antigen | GM-CSF |
|---|---|
| Clone | 6804 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| GM-CSF | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P04141 | |
| 1437 | |
| E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GM-CSF in ELISAs. In ELISAs, no cross-reactivity with recombinant mouse GM-CSF, recombinant rat GM-CSF, recombinant human (rh) M-CSF, rhG-CSF, rhIL-5-R alpha , or rhIL-3 R alpha is observed. |
| 6804 | |
| Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| CSF2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title