Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GM-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB215NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB215NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB215NA Supplier R&D Systems Supplier No. AB215NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GM-CSF Polyclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen GM-CSF
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-6 ug/mL
Gene Accession No. P04141
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Goat
Immunogen E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Test Specificity Detects human GM-CSF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) G-CSF, rhIL-1 alpha , rhIL-1 beta , rhIL-2, rhIL-3, rhIL-4, rhIL-6, rhTNF-alpha or rhTNF-beta is observed. Neutralizes the biological activity of both rhGM-CSF and natural human GM-CSF. It will not neutralize the biological activity of recombinant mouse GM-CSF.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.