Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GM-CSF R alpha Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7063U100UG
This item is not returnable.
View return policy
Description
GM-CSFR alpha Monoclonal antibody specifically detects GM-CSFR alpha in Human samples. It is validated for Flow CytometrySpecifications
GM-CSFR alpha | |
Monoclonal | |
Alexa Fluor 350 | |
P15509 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1438 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
IgG2b |
Flow Cytometry | |
1064847 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838 | |
Mouse myeloma cell line NS0-derived recombinant human GM-CSFRalpha, Met1-Gly320, Accession # P15509 | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction