Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GM-CSF R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB706
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB706 500 μg
MAB706SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB706 Supplier R&D Systems Supplier No. MAB706
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen GM-CSFR alpha
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 31916
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RA, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GMCSFR alpha, GM-CSFRa, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838
Gene Symbols CSF2RA
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GM-CSF R alpha Extracellular domain
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1438
Test Specificity Detects human GM-CSF R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) M-CSF R or rhGM-CSF R beta ( beta c ) is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.