Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GMF-beta Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.00
Specifications
Antigen | GMF-beta |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
GMF-beta Polyclonal specifically detects GMF-beta in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GMF-beta | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
glia maturation factor beta, glia maturation factor, beta, GMF, GMFB, GMFbeta, GMF-beta | |
GMFB | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P60983 | |
2764 | |
E. coli-derived recombinant human GMF-beta Met1-His142 Accession # P60983 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human GMF-beta in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title