Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human gp130 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
$561.50 - $585.00
Specifications
Antigen | gp130/CD130 |
---|---|
Concentration | LYOPH |
Applications | Western Blot, Neutralization |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
gp130/CD130 Polyclonal specifically detects gp130/CD130 in Human samples. It is validated for Western Blot, Neutralization.Specifications
gp130/CD130 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
P40189 | |
3572 | |
IgG | |
Affinity Purified |
LYOPH | |
Polyclonal | |
Goat | |
Human | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
IL6ST | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title