Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human gp130 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB628100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB628100 100 μg
MAB628SP 25 μg
MAB628500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB628100 Supplier R&D Systems Supplier No. MAB628100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen gp130/CD130
Applications Immunohistochemistry, Neutralization, ELISA
Classification Monoclonal
Clone 28105
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL, Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P40189
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Gene Symbols IL6ST
Host Species Mouse
Immunogen Sf 21-derived recombinant human gp130 extracellular domain Leu24-Glu619 (Glu619-Asp, predicted) Accession # P40189
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3572
Test Specificity Detects human gp130 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) CNTF, rhIL-6, rhIL-6 R, recombinant mouse (rm) IL-6, rhIL-11, rhLIF, rhLIF R, rmLIF, or rhOSM is observed. This antibody blocks the human gp130-mediated bioactivities induced by IL-6, IL-11, LIF, OSM, and CNTF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.