Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR101 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7495U100UG
This item is not returnable.
View return policy
Description
GPR101 Monoclonal specifically detects GPR101 in Human samples. It is validated for Flow Cytometry.Specifications
GPR101 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101 | |
NS0 mouse myeloma cell line transfected with human GPR101 Met1-Pro508, Accession # NP_473362 | |
RUO | |
83550 | |
Human | |
IgG |
Flow Cytometry | |
909603 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_473362 | |
Mouse | |
100 μg | |
Primary | |
Detects human GPR101 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction