Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR108 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.70 - $485.00
Specifications
Antigen | GPR108 |
---|---|
Clone | 1047612 |
Dilution | 1 μg/mL, 5 to 25 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11128100
![]() |
R&D Systems
MAB11128100 |
100 μg |
Each for $485.00
|
|
|||||
MAB11128SP
![]() |
R&D Systems
MAB11128SP |
25 μg |
Each for $188.70
|
|
|||||
Description
GPR108 Monoclonal antibody specifically detects GPR108 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
GPR108 | |
1 μg/mL, 5 to 25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 108, LUSTR2Lung seven transmembrane receptor 2, MGC14393, protein GPR108 | |
Human GPR108 synthetic peptide, Accession # Q9NPR9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1047612 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9NPR9 | |
56927 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title