Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR111 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6494V100UG
This item is not returnable.
View return policy
Description
GPR111 Monoclonal specifically detects GPR111 in Human samples. It is validated for Flow Cytometry.Specifications
GPR111 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
G protein-coupled receptor 111, PGR20 | |
Chinese Hamster Ovary cell line, CHO-derived recombinant human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
100 μg | |
222611 | |
Human | |
Supernatant |
Flow Cytometry | |
594519 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q8IZF7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human GPR111 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction