Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR111 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB64941SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB64941SP 25 μg
MAB64941100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB64941SP Supplier R&D Systems Supplier No. MAB64941SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR111 Monoclonal specifically detects GPR111 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen GPR111
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 594522
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q8IZF7
Gene Alias G protein-coupled receptor 111, PGR20
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222611
Test Specificity Detects human GPR111 in direct ELISAs. Stains human GPR111 HEK293-transfected cells and does not stain irrelevant non-GPR111 transfectants by Flow Cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.