Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR115 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5437
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5437 Supplier R&D Systems Supplier No. MAB5437
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR115 Monoclonal specifically detects GPR115 in Human samples. It is validated for Western Blot.

Specifications

Antigen GPR115
Applications Western Blot
Classification Monoclonal
Clone 527003
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IZF3
Gene Alias FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor
Gene Symbols GPR115
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GPR115 Ser22-Ala347 Accession # Q8IZF3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221393
Test Specificity Detects human GPR115 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human GPR30, 49, 56, 111, 114, 124, or 125 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.