Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR116 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000062
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000062 Supplier R&D Systems Supplier No. FAB112481U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR116 Monoclonal antibody specifically detects GPR116 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen GPR116
Applications Flow Cytometry
Classification Monoclonal
Clone 1055815
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8IZF2
Gene Alias FLJ90640, G protein-coupled receptor 116, Ig-Hepta homolog, KIAA0758DKFZp564O1923, KPG_001, probable G-protein coupled receptor 116
Host Species Mouse
Immunogen E. coli -expressed recombinant Human GPR116 extracellular domain., His643-Asn945, Accession # Q8IZF2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221395
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.