Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR15 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Supplier: R&D Systems MAB3654
This item is not returnable.
View return policy
Description
GPR15 Monoclonal specifically detects GPR15 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.Specifications
GPR15 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
P49685 | |
GPR15 | |
NS0 mouse myeloma cell line transfected with human GPR15 Met1-Leu360 Accession # P49685 | |
100 μg | |
Primary | |
Stains human GPR15-transfected cells but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
367902 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2838 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction