Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR22 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4500
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4500 Supplier R&D Systems Supplier No. MAB4500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR22 Monoclonal specifically detects GPR22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen GPR22
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 455108
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99680
Gene Alias G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22
Gene Symbols GPR22
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GPR22 Met1-Asp433 Accession # Q99680
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2845
Test Specificity Detects human GPR22. Stains human GPR22 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.