Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR26 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8449G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8449G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8449G100 Supplier R&D Systems Supplier No. FAB8449G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR26 Monoclonal specifically detects GPR26 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPR26
Applications Flow Cytometry
Classification Monoclonal
Clone 891318
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8NDV2
Gene Alias DKFZp761H2121, FLJ33774, G protein-coupled receptor 26, G-protein coupled receptor 26, MGC138216
Gene Symbols GPR26
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR26 Met1-Glu337 Accession # Q8NDV2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2849
Test Specificity Stains human GPR26 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.