Learn More
Human GPR26 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB8449N100UG
Catalog No. FAB8449N100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
GPR26 Monoclonal specifically detects GPR26 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR26
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8NDV2
GPR26
NS0 mouse myeloma cell line transfected with human GPR26 Met1-Glu337 Accession # Q8NDV2
RUO
2849
Human
IgG2b
Flow Cytometry
891318
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
DKFZp761H2121, FLJ33774, G protein-coupled receptor 26, G-protein coupled receptor 26, MGC138216
Mouse
100 μg
Primary
Stains human GPR26 transfectants but not irrelevant transfectants in flow cytometry.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)