Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR34 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4617G100UG
This item is not returnable.
View return policy
Description
GPR34 Monoclonal specifically detects GPR34 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
GPR34 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UPC5 | |
GPR34 | |
NS0 mouse myeloma cell line transfected with human GPR34 Accession # Q9UPC5 | |
RUO | |
2857 | |
Human | |
IgG2a |
Flow Cytometry | |
419859 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
G protein-coupled receptor 34, probable G-protein coupled receptor 34 | |
Mouse | |
100 μg | |
Primary | |
Stains human GPR34 transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction