Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR37 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4450G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4450G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4450G100 Supplier R&D Systems Supplier No. IC4450G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR37 Monoclonal specifically detects GPR37 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen GPR37
Applications Flow Cytometry
Classification Monoclonal
Clone 436923
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O15354
Gene Alias EDNRBL, Endothelin B receptor-like protein 1, ETBR-LP-1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELR, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37
Gene Symbols GPR37
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR37 Ala27-Cys613 Accession # O15354
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2861
Test Specificity Detects human GPR37.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.